EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Glycyrrhizic acid derivatives as influenza A/H1N1 virus inhibitors.
PMID 25801933
Abstract
This Letter describes the synthesis and antiviral activity study of some glycyrrhizic acid (GL) derivatives against influenza A/H1N1/pdm09 virus in MDCK cells. Conjugation of GL with l-amino acids or their methyl esters, and amino sugar (d-galactose amine) dramatically changed its activity. The most active compounds were GL conjugates with aromatic amino acids methyl esters (phenylalanine and tyrosine) (SI=61 and 38), and S-benzyl-cysteine (SI=71). Thus modification of GL is a perspective route in the search of new antivirals, and some of GL derivatives are potent as anti-influenza A/H1N1 agents.