EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Helicobacter pylori FlhA Binds the Sensor Kinase and Flagellar Gene Regulatory Protein FlgS with High Affinity.
PMID 25802298
Abstract
Flagellar biogenesis is a complex process that involves multiple checkpoints to coordinate transcription of flagellar genes with the assembly of the flagellum. In Helicobacter pylori, transcription of the genes needed in the middle stage of flagellar biogenesis is governed by RpoN and the two-component system consisting of the histidine kinase FlgS and response regulator FlgR. In response to an unknown signal, FlgS autophosphorylates and transfers the phosphate to FlgR, initiating transcription from RpoN-dependent promoters. In the present study, export apparatus protein FlhA was examined as a potential signal protein. Deletion of its N-terminal cytoplasmic sequence dramatically decreased expression of two RpoN-dependent genes, flaB and flgE. Optical biosensing demonstrated a high-affinity interaction between FlgS and a peptide consisting of residues 1 to 25 of FlhA (FlhANT). The KD (equilibrium dissociation constant) was 21 nM and was characterized by fast-on (kon = 2.9 × 10(4) M(-1)s(-1)) and slow-off (koff = 6.2 × 10(-4) s(-1)) kinetics. FlgS did not bind peptides consisting of smaller fragments of the FlhANT sequence. Analysis of binding to purified fragments of FlgS demonstrated that the C-terminal portion of the protein containing the kinase domain binds FlhANT. FlhANT binding did not stimulate FlgS autophosphorylation in vitro, suggesting that FlhA facilitates interactions between FlgS and other structures required to stimulate autophosphorylation. The high-affinity binding of FlgS to FlhA characterized in this study points to an additional role for FlhA in flagellar assembly. Beyond its necessity for type III secretion, the N-terminal cytoplasmic sequence of FlhA is required for RpoN-dependent gene expression via interaction with the C-terminal kinase domain of FlgS.