EMAIL THIS PAGE TO A FRIEND
International journal of medical microbiology : IJMM
Analysis of a single Helicobacter pylori strain over a 10-year period in a primate model.
PMID 25804332
Abstract
Helicobacter pylori from different individuals exhibits substantial genetic diversity. However, the kinetics of bacterial diversification after infection with a single strain is poorly understood. We investigated evolution of H. pylori following long-term infection in the primate stomach; Rhesus macaques were infected with H. pylori strain USU101 and then followed for 10 years. H. pylori was regularly cultured from biopsies, and single colony isolates were analyzed. At 1-year, DNA fingerprinting showed that all output isolates were identical to the input strain; however, at 5-years, different H. pylori fingerprints were observed. Microarray-based comparative genomic hybridization revealed that long term persistence of USU101 in the macaque stomach was associated with specific whole gene changes. Further detailed investigation showed that levels of the BabA protein were dramatically reduced within weeks of infection. The molecular mechanisms behind this reduction were shown to include phase variation and gene loss via intragenomic rearrangement, suggesting strong selective pressure against BabA expression in the macaque model. Notably, although there is apparently strong selective pressure against babA, babA is required for establishment of infection in this model as a strain in which babA was deleted was unable to colonize experimentally infected macaques.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6889
Famotidine
C8H15N7O2S3
T7883
Trimethoprim, ≥98% (HPLC)
C14H18N4O3
92131
Trimethoprim, crystallized, ≥99.0% (HPLC)
C14H18N4O3