EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Solid state compatibility study and characterization of a novel degradation product of tacrolimus in formulation.
PMID 25804434
Abstract
Tacrolimus is macrolide drug that is widely used as a potent immunosuppressant. In the present work compatibility testing was conducted on physical mixtures of tacrolimus with excipients and on compatibility mixtures prepared by the simulation of manufacturing process used for the final drug product preparation. Increase in one major degradation product was detected in the presence of magnesium stearate based upon UHPLC analysis. The degradation product was isolated by preparative HPLC and its structure was elucidated by NMR and MS studies. Mechanism of the formation of this degradation product is proposed based on complementary degradation studies in a solution and structural elucidation data. The structure was proven to be alpha-hydroxy acid which is formed from the parent tacrolimus molecule through a benzilic acid type rearrangement reaction in the presence of divalent metallic cations. Degradation is facilitated at higher pH values.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4705
Calcium acetate hydrate, BioXtra, ≥99% (titration)
C4H6CaO4 · xH2O
C1000
Calcium acetate hydrate, ReagentPlus®, ≥99% (titration), powder
C4H6CaO4 · xH2O
21056
Calcium acetate hydrate, BioUltra, ≥99.0% (calc. on dried material, KT)
C4H6CaO4 · xH2O
450146
Calcium hydroxide, 99.995% trace metals basis
H2CaO2
M2545
Magnesium acetate tetrahydrate, BioXtra, ≥99%
C4H6MgO4 · 4H2O
M5661
Magnesium acetate tetrahydrate, for molecular biology, ≥99%
C4H6MgO4 · 4H2O
CDS000001
Magnesium citrate, AldrichCPR
C6H6MgO7
529699
Magnesium oxide, ≥99.99% trace metals basis
MgO
203718
Magnesium oxide, 99.99% trace metals basis
MgO
549649
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
MgO
634646
Magnesium oxide, (single crystal substrate), ≥99.9% trace metals basis, <100>, L × W × thickness 10 mm × 10 mm × 0.5 mm
MgO
63138
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
MgO4S · 7H2O
RES0089M-A7
Magnesium sulfate heptahydrate, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
MgO4S · 7H2O
M5921
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
MgO4S · 7H2O
M2773
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
MgO4S · 7H2O
M1170
Met-Arg-Phe-Ala acetate salt, ≥90% (HPLC)
C23H37N7O5S