EMAIL THIS PAGE TO A FRIEND
Indian Journal of Microbiology
Molecular, Physiological and Phenotypic Characterization of Paracoccus denitrificans ATCC 19367 Mutant Strain P-87 Producing Improved Coenzyme Q10.
PMID 25805905
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9538
Coenzyme Q10, ≥98% (HPLC)
C59H90O4