PloS one
Anti-inflammatory activity of Choisya ternata Kunth essential oil, ternanthranin, and its two synthetic analogs (methyl and propyl N-methylanthranilates).
PMID 25807367
Abstract
Choisya ternata Kunth (Rutaceae) is native to North America where it is popularly known as "Mexican orange". In this study, the anti-inflammatory effects of the essential oil (EO) obtained from the leaves of C. ternata, one of its minor components (ternanthranin-ISOAN) and its two synthetic analogues (methyl and propyl N-methylanthranilate--MAN and PAN) were evaluated. Mice pretreated with the EO (EO) obtained from C. ternata leaves (3-100 mg/kg, p.o.), ISOAN, MAN or PAN (1-30 mg/kg, p.o.) and the reference drugs, morphine (1 mg/kg, p.o.) and acetylsalicylic acid (ASA, 100 mg/kg, p.o.), were evaluated in inflammation models such as formalin and subcutaneous air pouch models, with measurement of cell migration, exudate volume, protein extravasation, nitric oxide and pro-inflammatory cytokines. The EO from C. ternata significantly inhibited the time that the animals spent licking the formalin-injected paw in the second phase of the model at their higher doses (30 and 100 mg/kg, respectively). An inhibition of the inflammatory reaction induced after subcutaneous carrageenan injection into air pouch was also observed. In this model, the EO significantly reduced cell migration, exudate volume, protein extravased, and the increase in levels of inflammatory mediators (nitric oxide, TNF-α and IL-1β). ISOAN, MAN and PAN behaved in the same fashion at much smaller doses. Also, these molecules were able to show significant effects in the reduction of paw edema (at all tested doses) when the phlogistic agent was carrageenan, bradykinin, 5-HT, PGE2, C48/80 or 12-O-tetradecanoylphorbol-acetate (TPA). None of the tested doses had any effect in reducing histamine-induced edema. Our results indicate that the EO from C. ternata and anthranilate derivatives demonstrates an anti-inflammatory effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5376
Acetylsalicylic acid, ≥99.0%
C9H8O4
A3160
Acetylsalicylic acid, analytical standard
C9H8O4
V900185
Acetylsalicylic acid, Vetec, reagent grade, ≥99%
C9H8O4
A0200000
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
C9H8O4
Y0001460
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
C9H8O4
A2093
Aspirin, meets USP testing specifications
C9H8O4
1044006
Aspirin, United States Pharmacopeia (USP) Reference Standard
C9H8O4
PHR1003
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
C9H8O4
PHR1526
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
C22H29FO5
1176007
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
C22H29FO5
D0700000
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
46165
Dexamethasone, VETRANAL®, analytical standard
C22H29FO5
31375
Dexamethasone, tested according to Ph. Eur.
C22H29FO5
BP578
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
C22H29FO5
Y0001593
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
Y0001177
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
C22H29FO5
296082
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
C4H10O
91238
Diethyl ether, analytical standard
C4H10O
D2250000
Dinoprostone, European Pharmacopoeia (EP) Reference Standard
C20H32O5
47673
Formaldehyde solution, tested according to Ph. Eur.
CH2O
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
H7125
Histamine, ≥97.0%
C5H9N3
59964
Histamine, analytical standard
C5H9N3
793612
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
MgSO4
203726
Magnesium sulfate, ≥99.99% trace metals basis
MgSO4
P0409
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
C20H32O5
P5640
Prostaglandin E2, ≥93% (HPLC), synthetic
C20H32O5
P6532
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
C20H32O5