EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A Trigonal-Pyramidal Erbium(III) Single-Molecule Magnet.
PMID 25809280
Abstract
Given the recent advent of mononuclear single-molecule magnets (SMMs), a rational approach based on lanthanides with axially elongated f-electron charge cloud (prolate) has only recently received attention. We report herein a new SMM, [Li(THF)4[Er{N(SiMe3)2}3Cl]⋅2 THF, which exhibits slow relaxation of the magnetization under zero dc field with an effective barrier to the reversal of magnetization (ΔEeff/kB =63.3 K) and magnetic hysteresis up to 3 K at a magnetic field sweep rate of 34.6 Oe s(-1). This work questions the theory that oblate or prolate lanthanides must be stabilized with the appropriate ligand framework in order for SMM behavior to be favored.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219274
Eicosane, 99%
C20H42