EMAIL THIS PAGE TO A FRIEND
American journal of hematology
Pitfalls in the diagnosis of heparin-Induced thrombocytopenia: A 6-year experience from a reference laboratory.
PMID 25809312
Abstract
Heparin-induced thrombocytopenia (HIT) is caused by platelet-activating antibodies against complexes of platelet factor 4 (PF4) and heparin. The diagnosis of HIT is contingent on accurate and timely laboratory testing. Recently, alternative anticoagulants for the treatment of HIT have been introduced along with algorithms for better HIT diagnosis. However, the increased reliance on immunoassays for the diagnosis of HIT may have harmful consequences due to the high rate of false positive results. To compare trends and implications of current HIT testing approaches, we analyzed results over a six-year period from the McMaster University Platelet Immunology Reference Laboratory. From 2008 to 2013, 8,546 samples were investigated for HIT using both an in-house IgG-specific anti-PF4/heparin enzyme immunoassay (EIA) and the serotonin-release assay (SRA). Of 8,546 samples tested, 13.4% were true-positives (positive in both assays); 65.6% were true-negatives (negative in both assays); 20.9% were presumed false positive for HIT (EIA-positive/SRA-negative); and 0.2% were EIA-negative/SRA-positive. The frequency of EIA-positive/SRA-negative results increased over time (from 12.9% in 2008 to 22.9% in 2013). We found that the number of SRA-negative samples was reduced from referring centers that used an immunoassay as an initial screen; however, 41% of those samples tested negative in the immunoassay and in the SRA at the reference laboratory. The suspicion of HIT continues at a high rate and the agreement between the EIA and SRA test results remains problematic.