PloS one
Identification of multiple metabolic enzymes from mice cochleae tissue using a novel functional proteomics technology.
PMID 25811366
Abstract
A new type of technology in proteomics was developed in order to separate a complex protein mixture and analyze protein functions systematically. The technology combines the ability of two-dimensional gel electrophoresis (2-DE) to separate proteins with a protein elution plate (PEP) to recover active proteins for functional analysis and mass spectrometry (MS)-based identification. In order to demonstrate the feasibility of this functional proteomics approach, NADH and NADPH-dependent oxidases, major redox enzyme families, were identified from mice cochlear tissue after a specific drug treatment. By comparing the enzymatic activity between mice that were treated with a drug and a control group significant changes were observed. Using MS, five NADH-dependent oxidases were identified that showed highly altered enzymatic activities due to the drug treatment. In essence, the PEP technology allows for a systematic analysis of a large enzyme family from a complex proteome, providing insights in understanding the mechanism of drug treatment.