EMAIL THIS PAGE TO A FRIEND
Molecular imaging
Irbesartan attenuates atherosclerosis in Watanabe heritable hyperlipidemic rabbits: noninvasive imaging of inflammation by 18F-fluorodeoxyglucose positron emission tomography.
PMID 25812568
Abstract
The purpose of this study was to assess the usefulness of 18F-fluorodeoxyglucose positron emission tomography (18F-FDG PET) in evaluating the antiatherogenic effects of irbesartan, an angiotensin II type 1 receptor blocker. Watanabe heritable hyperlipidemic rabbits were divided into the irbesartan-treated group (75 mg/kg/d; n  =  14) and the control group (n  =  14). After a 9-month treatment, rabbits underwent 18F-FDG PET. Using the aortic lesions, autoradiography and histologic examinations were performed. PET imaging clearly visualized the thoracic lesions of control rabbits and showed a significant decrease in the 18F-FDG uptake level of irbesartan-treated rabbits (78.8% of controls; p < .05). Irbesartan treatment significantly reduced the plaque size (43.1% of controls) and intraplaque macrophage infiltration level (48.1% of controls). The 18F-FDG uptake level in plaques positively correlated with the plaque size (r  =  .65, p < .05) and macrophage infiltration level (r  =  .57, p < .05). Noninvasive imaging by 18F-FDG PET is useful for evaluating the therapeutic effects of irbesartan and reflects inflammation, a key factor involved in the therapeutic effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I2286
Irbesartan, ≥98% (HPLC), powder
C25H28N6O