EMAIL THIS PAGE TO A FRIEND
Neurobiology of Disease
Potential role of adolescent alcohol exposure-induced amygdaloid histone modifications in anxiety and alcohol intake during adulthood.
PMID 25814047