PLoS ONE
Actin-mediated gene expression depends on RhoA and Rac1 signaling in proximal tubular epithelial cells.
PMID 25816094
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C106
CGS-12066 maleate salt, solid
C17H17F3N4 · C4H4O4
03609
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
C10H16N2O8
1295607
Glycerin, United States Pharmacopeia (USP) Reference Standard
C3H8O3
PHR1020
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
C3H8O3
49779
Glycerol, tested according to Ph.Eur., anhydrous
C3H8O3
77067
Glycerol, analytical standard
C3H8O3
12146
Glycerol 3 M solution, 3 M
C3H8O3 · xH2O
54461
HEPES, ≥99.0% (T)
C8H18N2O4S
PHR1428
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
C8H18N2O4S
1316004
Hydrocortisone, United States Pharmacopeia (USP) Reference Standard
C21H30O5
PHR1014
Hydrocortisone, Pharmaceutical Secondary Standard; Certified Reference Material
C21H30O5
31719
Hydrocortisone, VETRANAL, analytical standard
C21H30O5
H1300000
Hydrocortisone, European Pharmacopoeia (EP) Reference Standard
C21H30O5
BP576
Hydrocortisone, British Pharmacopoeia (BP) Assay Standard
C21H30O5
Y0001092
Hydrocortisone for peak identification, European Pharmacopoeia (EP) Reference Standard
C21H30O5
T0198
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone D66, purified from hybridoma cell culture
71383
Sodium chloride, tested according to Ph.Eur.
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, according to ISO 17025, ≥99.5%
ClNa
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71177
Sodium standard for AAS, ready-to-use, traceable to BAM, in H2O
ClNa
Y0503
Y-27632 dihydrochloride, ≥98% (HPLC)
C14H21N3O · 2HCl