EMAIL THIS PAGE TO A FRIEND
Nature communications
Cmr1/WDR76 defines a nuclear genotoxic stress body linking genome integrity and protein quality control.
PMID 25817432
Abstract
DNA replication stress is a source of genomic instability. Here we identify changed mutation rate 1 (Cmr1) as a factor involved in the response to DNA replication stress in Saccharomyces cerevisiae and show that Cmr1--together with Mrc1/Claspin, Pph3, the chaperonin containing TCP1 (CCT) and 25 other proteins--define a novel intranuclear quality control compartment (INQ) that sequesters misfolded, ubiquitylated and sumoylated proteins in response to genotoxic stress. The diversity of proteins that localize to INQ indicates that other biological processes such as cell cycle progression, chromatin and mitotic spindle organization may also be regulated through INQ. Similar to Cmr1, its human orthologue WDR76 responds to proteasome inhibition and DNA damage by relocalizing to nuclear foci and physically associating with CCT, suggesting an evolutionarily conserved biological function. We propose that Cmr1/WDR76 plays a role in the recovery from genotoxic stress through regulation of the turnover of sumoylated and phosphorylated proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N8141
4-Nitroquinoline N-oxide, ≥98%
C9H6N2O3
A8626
Adenine, ≥99%
C5H5N5
A2786
Adenine, BioReagent, suitable for cell culture
C5H5N5
A5665
Adenine, BioReagent, suitable for plant cell culture, ≥99%
C5H5N5
PHR1383
Adenine, Pharmaceutical Secondary Standard; Certified Reference Material
C5H5N5
A0230000
Adenine, European Pharmacopoeia (EP) Reference Standard
C5H5N5
HPA040626
Anti-WDR76 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
03620
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
C10H16N2O8
03609
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
C10H16N2O8
03690
Ethylenediaminetetraacetic acid solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
C10H16N2O8
47072
Fluoride ion solution for ISE, 0.1 M F-, analytical standard (for ion-selective electrodes)
FNa
80373
Fluoride, certified anion standard solution, 1000 mg/kg F-, for ion chromatography, certified by BAM, according to ISO 17025
FNa
A5006
L-Arginine, reagent grade, ≥98%
C6H14N4O2
A8094
L-Arginine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C6H14N4O2
A4474
L-Arginine, PharmaGrade, Ajinomoto, EP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C6H14N4O2
11009
L-Arginine, BioUltra, ≥99.5% (NT)
C6H14N4O2
W381918
L-Arginine, 99%, FCC, FG
C6H14N4O2
PHR1106
L-Arginine, Pharmaceutical Secondary Standard; Certified Reference Material
C6H14N4O2
L5501
L-Lysine, ≥98% (TLC)
C6H14N2O2
62840
L-Lysine, crystallized, ≥98.0% (NT)
C6H14N2O2
23128
L-Lysine, analytical standard
C6H14N2O2
62850
L-Lysine solution, purum, 50% in H2O
C6H14N2O2
608041
L-Lysine-13C6,15N2 hydrochloride, 99 atom % 15N, 99 atom % 13C, 95% (CP)
13C6H15Cl15N2O2
PHR1321
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
ClNa
71387
Sodium chloride, certified reference material for titrimetry, certified by BAM, ≥99.5%
ClNa
71383
Sodium chloride, tested according to Ph Eur
ClNa
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa
PHR1408
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
FNa
67414
Sodium fluoride 0.5 M solution
FNa