EMAIL THIS PAGE TO A FRIEND
Journal of Chromatography A
A new anion-exchange/hydrophobic monolith as stationary phase for nano liquid chromatography of small organic molecules and inorganic anions.
PMID 25817709