EMAIL THIS PAGE TO A FRIEND
Biomaterials
Development and functional evaluation of biomimetic silicone surfaces with hierarchical micro/nano-topographical features demonstrates favourable in vitro foreign body response of breast-derived fibroblasts.
PMID 25818416