EMAIL THIS PAGE TO A FRIEND
Journal of Crohn's & colitis
Changes in Epithelial Barrier Function in Response to Parasitic Infection: Implications for IBD Pathogenesis.
PMID 25820018
Abstract
Mast cells [MCs] are implicated in epithelial barrier alterations that characterize inflammatory and functional bowel disorders. In this study, we describe mast cell proteinases [chymases and tryptases] and tight junction [TJ] proteins kinetics in a rat model of postinfectious gut dysfunction. Jejunal tissues of control and -infected rats were used. Inflammation-related changes in MCs and the expression of TJ-related proteins were evaluated by immunostaining and reverse transcription-quantitative polymerase chain reaction. Epithelial barrier function was assessed in vitro (Ussing chambers) and in vivo. After infection, intestinal inflammation was associated with a generalized overexpression of MC chymases, peaking between Days 6 and 14. Thereafter, a mucosal MC hyperplasia and a late increase in connective tissue MC counts were observed. From Day 2 post-infection, TJ proteins occludin and claudin-3 expression was down-regulated whereas the pore-forming protein claudin-2 was overexpressed. The expression of proglucagon, precursor of the barrier-enhancing factor glucagon-like peptide-2, was reduced. These changes were associated with an increase in epithelial permeability, both in vitro and in vivo. Proteinases expression and location of mucosal and connective tissue MCs indicate a time-related pattern in the maturation of intestinal MCs following infection. Altered expression of TJ-related proteins is consistent with a loss of epithelial tightness, and provides a molecular mechanism for the enhanced epithelial permeability observed in inflammatory conditions of the gut.