EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Carbamates: A study on genotoxic, cytotoxic, and apoptotic effects induced in Chinese hamster ovary (CHO-K1) cells.
PMID 25820133
Abstract
In vitro effects of the carbamates pirimicarb and zineb and their formulations Aficida® (50% pirimicarb) and Azzurro® (70% zineb), respectively, were evaluated in Chinese hamster ovary (CHO-K1) cells. Whereas the cytokinesis-blocked micronucleus cytome assay was employed to test for genotoxicity, MTT, neutral red (NR), and apoptosis evaluation were used as tests for estimating cell viability and succinic dehydrogenase activity, respectively. Concentrations tested were 10-300 μg/ml for pirimicarb and Aficida®, and 1-50 μg/ml for zineb and Azzurro®. All compounds were able to increase the frequency of micronuclei. A marked reduction in the nuclear division index was observed after treatment with 5 μg/ml of zineb and Azzurro® and 10 μg/ml of Azzurro®. Alterations in the cellular morphology not allowing the recognition of binucleated cells exposed to 300 μg/ml pirimicarb and Aficida® as well as 10-50 μg/ml zineb and Azzurro®. All four compounds induced inhibition of both cell viability and succinic dehydrogenase activity and trigger apoptosis in CHO-K1 cells, at least when exposed for 24 h. The data herein demonstrate the genotoxic and cytotoxic effects exerted by these carbamates and reveal the potential risk factor of these pesticides, still extensively used worldwide, for both human health and the environment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E-034
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-040
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material
C2H6O
E-031
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-038
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-041
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-056
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-043
Ethanol-20 (5 ampules/kit), 20 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-032
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-039
Ethanol-200 (5 ampules/kit), 200 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-035
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-033
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-045
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-036
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-044
Ethanol-400 (5 ampules/kit), 400 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
E-029
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-053
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-030
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material
C2H6O
E-037
Ethanol-80, 80 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material
C2H6O
793175
Reagent Alcohol, ACS reagent
793183
Reagent Alcohol, 95%
793191
Reagent Alcohol, 80%
793213
Reagent Alcohol, 70%
277649
Reagent Alcohol, anhydrous, ≤0.003% water
676829
Reagent Alcohol, anhydrous, ≤0.005% water
R8382
Reagent Alcohol, used for histology tissue preparation
R3404
Reagent Alcohol, 95%, Used for histology tissue preparation
R3279
Reagent Alcohol, 80%, used for histology tissue preparation
R3154
Reagent Alcohol, 70%, used for histology tissue preparation
362808
Reagent Alcohol, reagent grade