EMAIL THIS PAGE TO A FRIEND
The American journal of medicine
Impact of a clinical pharmacist stress ulcer prophylaxis management program on inappropriate use in hospitalized patients.
PMID 25820164
Abstract
Appropriate utilization of stress ulcer prophylaxis should be limited to high-risk, intensive care unit (ICU) patients. However, inappropriate stress ulcer prophylaxis use among all hospitalized patients remains a concern. The purpose of this study was to evaluate the clinical and economic impact of a novel pharmacist-managed stress ulcer prophylaxis program in ICU and general ward patients. This retrospective, pre- and poststudy design was conducted in adult ICU and general ward patients at a large academic medical center between January 1, 2011 and January 31, 2012 to compare the rates of inappropriate stress ulcer prophylaxis before and after the implementation of a pharmacist-led stress ulcer prophylaxis management program. A total of 1134 unique patients consisting of 16,415 patient days were evaluated. The relative reduction in the rate of inappropriate stress ulcer prophylaxis days after program implementation in ICU and general ward patients was 58.3% and 83.5%, respectively (P < .001). The rates of ICU patients inappropriately continued on stress ulcer prophylaxis upon hospital discharge in the pre- and postimplementation groups were 29.9% and 3.6%, respectively (P < .001), whereas general ward patients significantly decreased from 36.2% to 5.4% in the pre- and postimplementation groups, respectively (Pxa0 $200,000 annually. No differences in clinical outcomes were observed. The implementation of a pharmacist-managed stress ulcer prophylaxis program was associated with a decrease in inappropriate acid suppression rates during hospitalization and upon discharge, as well as significant cost savings.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6889
Famotidine
C8H15N7O2S3
L8533
Lansoprazole, ≥98% (TLC), powder
C16H14F3N3O2S