EMAIL THIS PAGE TO A FRIEND
Hippocampus
Orexinergic theta rhythm in the rat hippocampal formation: In vitro and in vivo findings.
PMID 25820995
Abstract
Previous in vivo data suggested that orexin neuropeptides (ORX(A) and ORX(B) ) synthetized in hypothalamic neurons were involved in the mechanism of generation of the hippocampal formation theta rhythm. Surprisingly, this suggestion has never been directly proved by experiments using intraseptal or intrahippocampal administration of orexins. In this study, involving the use of in vitro hippocampal formation slices and in vivo model of anesthetized rat, we provide the first convergent electropharmacological evidence that in the presence of both ORX(A) and ORX(B) the hippocampal formation neuronal network is capable of producing oscillations in the theta band. This effect of orexin peptides was antagonized by selective blockers of orexin receptors (OX1 R and OX2 R), SB 334867 and TCS OX2 29, respectively. These results provide evidence for a novel, orexinergic mechanism responsible for the production of theta rhythm in the hippocampal formation neuronal network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4388
2-Bromo-2-chloro-1,1,1-trifluoroethane, ≥99%
C2HBrClF3