EMAIL THIS PAGE TO A FRIEND
The Journal of membrane biology
Transport of hydrophobic ions in erythrocyte membrane: I. Zero membrane potential properties.
PMID 2582132
Abstract
The permeability and partition coefficients of tetraphenylarsonium (TPA) and several other organic cations were studied in the human erythrocyte using an ion-selective electrode. The permeability constant for the different cations could be explained quite well by differences in oil/water partition coefficients. No evidence for facilitated transport could be found. Binding of the organic ions occurred to both the cell membrane and to intracellular contents. Partitioning to the membrane remained relatively constant despite variation from ion intracellular binding with blood samples from different donors. TPA flux is stimulated by substoichiometric amounts of tetraphenylboron and other organic anions, suggesting an ion-pairing mechanism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T25305
Tetraphenylarsonium(V) chloride hydrate, 97%
C24H20AsCl · xH2O