EMAIL THIS PAGE TO A FRIEND
Archiv der Pharmazie
Synthesis of carbamide derivatives bearing tetrahydroisoquinoline moieties and biological evaluation as analgesia drugs in mice.
PMID 25824579
Abstract
Transient receptor potential vanilloid 1 (TRPV1) is a ligand-gated non-selective cation channel that is considered to be an important pain integrator. Tetrahydroisoquinoline, the prototypical antagonist of TRPV1, has a clear therapeutic potential. Here, a series of carbamide derivatives of tetrahydroisoquinoline were designed and synthesized. Preliminary biological tests suggested that the compounds I 1, I 2, and I 9 had favorable TRPV1 antagonism activity. In further studies, I 1 exhibited better antinociceptive activity than the positive control BCTC in diverse pain models. All of these results suggested that I 1 can be considered as the lead candidate for the further development of antinociceptive drugs.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T13005
1,2,3,4-Tetrahydroisoquinoline, 95%
C9H11N
C2230
Coelenterazine, native, solid
C26H21N3O3