EMAIL THIS PAGE TO A FRIEND
Nature Communications
Glycosylation of immunoglobulin G determines osteoclast differentiation and bone loss.
PMID 25825024
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
09972
3-Nitrotetrazolium blue chloride, ≥75% (AT)
C40H30Cl2N10O6
50553
α-tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Ca3O8P2
09830
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
CH5NO3
16483
β-tri-Calcium phosphate, puriss. p.a., ≥95% β-phase basis (sintered powder)
Ca3O8P2
C8271
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
C20H30N4NaO16P
G0750
D-(+)-Galactose, ≥99%
C6H12O6
G0625
D-(+)-Galactose, ≥98%
C6H12O6
G6404
D-(+)-Galactose, BioXtra, pH 5.0-7.0 (20 °C, 1 M in H2O), ≥99%
C6H12O6
G5388
D-(+)-Galactose, powder, anhydrous, BioReagent, suitable for cell culture, suitable for insect cell culture
C6H12O6
48260
D-(+)-Galactose, for microbiology, ≥99.0%
C6H12O6
15522
D-(+)-Galactose, meets analytical specification of Ph. Eur., BP
C6H12O6
ARK2181
D-(+)-Mannose, PharmaGrade, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C6H12O6
M2069
D-(+)-Mannose, from wood, ≥99%
C6H12O6
M8574
D-(+)-Mannose, synthetic, ≥99%
C6H12O6
M6020
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
C6H12O6
63579
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
C6H12O6
63580
D-(+)-Mannose, for microbiology, ≥99%
C6H12O6
63582
D-(+)-Mannose, for microbiology, ≥99.0% (sum of enantiomers, HPLC)
C6H12O6
M4625
D-(+)-Mannose
C6H12O6
1287700 Galactose, United States Pharmacopeia (USP) Reference Standard
C6H12O6
PHR1206
Galactose, Pharmaceutical Secondary Standard; Certified Reference Material
C6H12O6
G0050000
Galactose, European Pharmacopoeia (EP) Reference Standard
C6H12O6
ARK2176
Galactose, plant-derived, PharmaGrade, USP/NF, Ph Eur, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H12O6
69947
MOPS, BioUltra, for molecular biology, ≥99.5% (T)
C7H15NO4S
T4375
Tetrazolium Blue Chloride, used in colorimetric determination of reducing compounds
C40H32Cl2N8O2
88190
Tetrazolium Blue Chloride, indicator for germination, for microbiology, ≥90% (T)
C40H32Cl2N8O2