EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Development of direct conversion method for microalgal biodiesel production using wet biomass of Nannochloropsis salina.
PMID 25827362
Abstract
In this work, the effects of several factors, such as temperature, reaction time, and solvent and acid quantity on in situ transesterification yield of wet Nannochloropsis salina were investigated. Under equivalent total solvent volume to biomass ratio, pure alcohol showed higher yield compared to alcohol-chloroform solvent. For esterifying 200 mg of wet cells, 2 ml of methanol and 1 ml of ethanol was sufficient to complete in situ transesterification. Under temperatures of 105 °C or higher, 2.5% and 5% concentrations of sulfuric acid was able to successfully convert more than 90% of lipid within 30 min when methanol and ethanol was used as solvents respectively. Also, it was verified that the optimal condition found in small-scale experiments is applicable to larger scale using 2 L scale reactor as well.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219274
Eicosane, 99%
C20H42