EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
The coreceptor CD4 is expressed in distinct nanoclusters and does not colocalize with T-cell receptor and active protein tyrosine kinase p56lck.
PMID 25829544