EMAIL THIS PAGE TO A FRIEND
Journal of Food Science and Technology
Evolution of antioxidants in dietary fiber powder produced from white cabbage outer leaves: effects of blanching and drying methods.
PMID 25829610