EMAIL THIS PAGE TO A FRIEND
Biopreservation and biobanking
Method validation for automated isolation of viable peripheral blood mononuclear cells.
PMID 25830476
Abstract
This article is part of a series of publications providing formal method validation for biospecimen processing in the context of accreditation in laboratories and biobanks. We report the optimization and validation for fitness-for-purpose of automated and manual protocols for isolating peripheral blood mononuclear cells (PBMCs) from whole blood, and compare the two methods. The manual method was optimized for whole blood centrifugation speed, gradient type (Ficoll, Leucosep, CPT), and freezing method (Mr Frosty, Controlled Rate Freezing). Various parameters of the automated protocol using a CPT gradient on a Tecan liquid handler were optimized. Optimal protocols were validated in parallel for reproducibility and robustness. Optimization and validation were assessed in terms of cell yield, viability, recovery, white blood cell (WBC) subpopulation distribution, gene expression, and lymphoblastoid cell line (LCL) transformation. An initial centrifugation of whole blood at 2000 g was considered optimal for further processing, allowing isolation of plasma and PBMCs from a single sample. The three gradients gave similar outcomes in terms of cell yield, viability, and WBC subpopulation distribution. Ficoll showed some advantages and was selected for further evaluations. Optimization of the automated protocol script using a CPT gradient gave 61% cell recovery. No significant differences in quality, quantity, and WBC subpopulation distribution were seen between the two freezing methods, and Mr. Frosty was selected. The manual and automated protocols were reproducible in terms of quantity, recovery, viability, WBC subpopulation distribution, gene expression, and LCL transformation. Most (75%-100%) of the 13 robustness parameters were accepted for both methods with an 8 h pre-centrifugation delay versus 38%-85% after 24 h. Differences identified between the automated and manual methods were not considered consequential. We validated the first fully automated method for isolating viable PBMCs, including RNA analysis and generation of LCLs. We recommend processing within 8 h of blood collection.