EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Prediction of the stability of meropenem in intravenous mixtures.
PMID 25832020
Abstract
The purpose of this study was to predict the stability of meropenem in a mixed infusion. The hydrolysis of meropenem in aqueous solution was found to be accelerated by pH, and by increasing concentrations of sodium bisulfite (SBS) and L-cysteine. Equations were derived for the degradation rate constants (kobs) of pH, SBS and L-cysteine, and fractional rate constants were estimated by the nonlinear least-squares method (quasi-Newton method using the solver in Microsoft Excel) at 25°C. The activation energy (Ea) and frequency factor (A) were calculated using the Arrhenius equation. The pH of the mixed infusion was estimated using the characteristic pH curve. From these results, an equation was derived giving the residual ratio (%) of meropenem at any time after mixing an infusion containing SBS and/or L-cysteine at any temperature, and in the pH range 4.0-10.0. A high correlation was shown to exist between the estimated and determined residual ratios (%).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1392454
Meropenem, United States Pharmacopeia (USP) Reference Standard
C17H25N3O5S · 3H2O