EMAIL THIS PAGE TO A FRIEND
Acta Pharmacologica Sinica
2-(3-Benzoylthioureido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic acid ameliorates metabolic disorders in high-fat diet-fed mice.
PMID 25832429
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2537
Mevastatin, ≥95% (HPLC), powder
C23H34O5
442785
Squalene, analytical standard
C30H50