EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Brominated polyunsaturated lipids with protein tyrosine phosphatase-1B inhibitory activity from Chinese marine sponge Xestospongia testudinaria.
PMID 25832997
Abstract
A new brominated polyunsaturated lipid, methyl (E,E)-14,14-dibromo-4,6,13-tetradecatrienoate (1), along with three known related analogues (2-4), were isolated from the Et2O-soluble portion of the acetone extract of Chinese marine sponge Xestospongia testudinaria treated with diazomethane. The structure of the new compound was elucidated by detailed spectroscopic analysis and by comparison with literature data. Compound 3 exhibited significant inhibitory activity against protein tyrosine phosphatase 1B (PTP1B), a key target for the treatment of type II diabetes and obesity, with an IC50 value of 5.30 ± 0.61 μM, when compared to the positive control oleanolic acid (IC50 = 2.39 ± 0.26 μM).