EMAIL THIS PAGE TO A FRIEND
Scandinavian journal of immunology
Chlamydia pneumoniae and Chlamydia Trachomatis Infection Differentially Modulates Human Dendritic Cell Line (MUTZ) Differentiation and Activation.
PMID 25833314
Abstract
Chlamydia trachomatis and Chlamydia pneumoniae are important human pathogens that infect the urogenital/anorectal and respiratory tracts, respectively. Whilst the ability of these bacteria to infect epithelia is well defined, there is also considerable evidence of infection of leucocytes, including dendritic cells (DCs). Using a human dendritic cell line (MUTZ), we demonstrate that the infection and replication of chlamydiae inside DCs is species and serovar specific and that live infection with C. pneumoniae is required to upregulate costimulatory markers CD80, CD83 and human leucocyte antigen (HLA)-DR on MUTZ cells, as well as induce secretion of interleukin (IL)-2, IL-6, IL-8, IL-12 (p70), interferon-gamma and tumour necrosis factor-alpha Conversely, C. trachomatis serovar D failed to upregulate DC costimulatory markers, but did induce secretion of high concentrations of IL-8. Interestingly, we also observed that infection of MUTZ cells with C. pneumoniae or C. trachomatis serovar L2, whilst not replicative, remained infectious and upregulated lymph node migratory marker CCR7 mRNA. Taken together, these data confirm the findings of other groups using primary DCs and demonstrate the utility of MUTZ cells for further studies of chlamydial infection.