EMAIL THIS PAGE TO A FRIEND
Development (Cambridge, England)
The loss of Hh responsiveness by a non-ciliary Gli2 variant.
PMID 25834022
Abstract
Hedgehog signaling is crucial for vertebrate development and physiology. Gli2, the primary effector of Hedgehog signaling, localizes to the tip of the primary cilium, but the importance of its ciliary localization remains unclear. We address the roles of Gli2 ciliary localization by replacing endogenous Gli2 with Gli2(ΔCLR), a Gli2 variant not localizing to the cilium. The resulting Gli2(ΔCLRKI) and Gli2(ΔCLRKI);Gli3 double mutants resemble Gli2-null and Gli2;Gli3 double mutants, respectively, suggesting the lack of Gli2(ΔCLR) activation in development. Significantly, Gli2(ΔCLR) cannot be activated either by pharmacochemical activation of Smo in vitro or by loss of Ptch1 in vivo. Finally, Gli2(ΔCLR) exhibits strong transcriptional activator activity in the absence of Sufu, suggesting that the lack of its activation in vivo results from a specific failure in relieving the inhibitory function of Sufu. Our results provide strong evidence that the ciliary localization of Gli2 is crucial for cilium-dependent activation of Hedgehog signaling.