EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Interaction of thrombospondin1 and CD36 contributes to obesity-associated podocytopathy.
PMID 25835637
Abstract
Obesity is associated with podocyte injury and the development of proteinuria. Elevated plasma free fatty acid is one of the characteristics of obesity and has been linked to podocyte dysfunction. However, the mechanisms remain unclear. In the current study, we examined the effect of saturated free fatty acid (FFA) on human podocyte apoptosis and function in vitro. The mechanism and its in vivo relevance were also determined. We found that FFA treatment induced human podocyte apoptosis and dysfunction, which was associated with increased expression of a matricellular protein-thrombospondin1 (TSP1). FFA stimulated TSP1 expression in podocytes at the transcriptional levels through activation of MAPK pathway. Addition of purified TSP1 to cell culture media induced podocyte apoptosis and dysfunction. Tis effect is though a TGF-β independent mechanism. Moreover, peptide treatment to block TSP1 binding to its receptor-CD36 attenuated FFA induced podocyte apoptosis, suggesting that TSP1/CD36 interaction mediates FFA-induced podocyte apoptosis. Importantly, using a diet-induced obese mouse model, in vivo data demonstrated that obesity-associated podocyte apoptosis and dysfunction were attenuated in TSP1 deficient mice as well as in CD36 deficient mice. Taken together, these studies provide novel evidence that the interaction of TSP1 with its receptor CD36 contributes to obesity--associated podocytopathy.