EMAIL THIS PAGE TO A FRIEND
BioMed research international
Synthesis and characterization of biodegradable polyurethane for hypopharyngeal tissue engineering.
PMID 25839041
Abstract
Biodegradable crosslinked polyurethane (cPU) was synthesized using polyethylene glycol (PEG), L-lactide (L-LA), and hexamethylene diisocyanate (HDI), with iron acetylacetonate (Fe(acac)3) as the catalyst and PEG as the extender. Chemical components of the obtained polymers were characterized by FTIR spectroscopy, (1)H NMR spectra, and Gel Permeation Chromatography (GPC). The thermodynamic properties, mechanical behaviors, surface hydrophilicity, degradability, and cytotoxicity were tested via differential scanning calorimetry (DSC), tensile tests, contact angle measurements, and cell culture. The results show that the synthesized cPU possessed good flexibility with quite low glass transition temperature (T g , -22°C) and good wettability. Water uptake measured as high as 229.7 ± 18.7%. These properties make cPU a good candidate material for engineering soft tissues such as the hypopharynx. In vitro and in vivo tests showed that cPU has the ability to support the growth of human hypopharyngeal fibroblasts and angiogenesis was observed around cPU after it was implanted subcutaneously in SD rats.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1012772
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
C2H6O
PHR1070
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
C2H6O
270997
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
439223
Dichloromethane, for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
CH2Cl2
154792
Dichloromethane, ACS spectrophotometric grade, ≥99.5%, contains 50-150 ppm amylene as stabilizer
CH2Cl2
270563
Dichloromethane, ≥99.9%
CH2Cl2
66742
Dichloromethane, Selectophore, ≥99.5%
CH2Cl2
02575
Dichloromethane, analytical standard
CH2Cl2
48538
Dichloromethane, analytical standard, stabilized
CH2Cl2
48600
Dichloromethane solution, certified reference material, 200 μg/mL in methanol
CH2Cl2
40042
Dichloromethane solution, certified reference material, 5000 μg/mL in methanol
CH2Cl2
723851
Dichloromethane solution, contains 10 % (v/v) methanol
CH2Cl2
270989
Ethyl acetate, anhydrous, 99.8%
C4H8O2
34858
Ethyl acetate, for HPLC, ≥99.7%
C4H8O2
437549
Ethyl acetate, ACS reagent, ≥99.5%
C4H8O2
58958
Ethyl acetate, analytical standard
C4H8O2
17023
Ethyl acetate, tested according to Ph Eur
C4H8O2
270520
Ethyl acetate, HPLC grade, 99.8%
C4H8O2
W241407
Ethyl acetate, ≥99%, FCC, FG
C4H8O2
W241415
Ethyl acetate, natural, ≥99%, FCC, FG
C4H8O2
537446
Ethyl acetate, ReagentPlus®, ≥99.8%
C4H8O2
L092004
Ethyl acetate
C4H8O2
L092003
Ethyl acetate
C4H8O2
PHR1481
Ethyl Acetate
C4H8O2
517003
Iron(III) acetylacetonate, ≥99.9% trace metals basis
C15H21FeO6
44920
Iron(III) acetylacetonate, purum, ≥97.0% (RT)
C15H21FeO6
F300
Iron(III) acetylacetonate, 97%
C15H21FeO6
M1550000
Methylene chloride, European Pharmacopoeia (EP) Reference Standard
CH2Cl2
PHR1557
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
CH2Cl2