EMAIL THIS PAGE TO A FRIEND
Journal of the American College of Surgeons
RETRACTED: How does high-concentration supplemental perioperative oxygen influence surgical outcomes after thyroid surgery? A prospective, randomized, double-blind, controlled, monocentric trial.
PMID 25840543
Abstract
This article has been retracted: please see Elsevier Policy on Article Withdrawal (https://www.elsevier.com/about/our-business/policies/article-withdrawal). This article has been retracted at the request of the Editor-in-Chief. An independent statistical analysis based on the summary data tables and statistical results reported in the article confirmed that the statistical results are incorrect and the data do not support the conclusions of the article.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1044800
Atracurium besylate, United States Pharmacopeia (USP) Reference Standard
C65H82N2O18S2
Y0000424
Atracurium besylate, European Pharmacopoeia (EP) Reference Standard
C65H82N2O18S2
Y0000503
Atracurium for impurity F identification, European Pharmacopoeia (EP) Reference Standard
C65H82N2O18S2
Y0000504
Atracurium for peak identification, European Pharmacopoeia (EP) Reference Standard
C65H82N2O18S2
1045009
Atropine sulfate, United States Pharmacopeia (USP) Reference Standard
C34H46N2O6 · H2O4S · H2O
A1400000
Atropine sulfate, European Pharmacopoeia (EP) Reference Standard
C34H46N2O6 · H2O4S · H2O
PHR1379
Atropine Sulfate, Pharmaceutical Secondary Standard; Certified Reference Material
C34H46N2O6 · H2O4S · H2O
A0257
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
C34H46N2O6 · H2O4S · H2O
Y0000797
Promethazine for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H20N2S · HCl
P4651
Promethazine hydrochloride
C17H20N2S · HCl
P3400000
Promethazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C17H20N2S · HCl
1570009
Promethazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C17H20N2S · HCl
46682
Promethazine hydrochloride, VETRANAL®, analytical standard
C17H20N2S · HCl
PHR1467
Promethazine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C17H20N2S · HCl