EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
A fast way to track functional OmpF reconstitution in liposomes: Escherichia coli total lipid extract.
PMID 25841673
Abstract
A major requirement to perform structural studies with membrane proteins is to define efficient reconstitution protocols that ensure a high incorporation degree and protein directionality and topology that mimics its in vivo conditions. For this kind of studies, protein reconstitution in membrane systems via a detergent-mediated pathway is usually successfully adopted because detergents are generally used in the initial isolation and purification of membrane proteins. This study reports OmpF reconstitution in preformed Escherichia coli liposomes followed by detection of its insertion by analyzing modifications on membrane structure by two different techniques: steady-state fluorescence anisotropy and dynamic light scattering. Another important issue is protein directionality. For OmpF, it is known that interaction with polyamines promotes channel blockage. In this work, the spermine-OmpF interaction was evaluated using surface plasmon resonance, and protein directionality was confirmed.