EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Release behavior of non-network proteins and its relationship to the structure of heat-induced soy protein gels.
PMID 25842998
Abstract
Heat-induced soy protein gels were prepared by heating protein solutions at 12%, 15% ,or 18% for 0.5, 1.0, or 2.0 h. The release of non-network proteins from gel slices was conducted in 10 mM pH 7.0 sodium phosphate buffer. SDS-PAGE and diagonal electrophoresis demonstrated that the released proteins consisted of undenatured AB subunits and denatured proteins including monomers of A polypeptides, disulfide bond linked dimers, trimers, and polymers of A polypeptides, and an unidentified 15 kDa protein. SEC-HPLC analysis of non-network proteins revealed three major protein peaks, with molecular weights of approximately 253.9, 44.8, and 9.7 kDa. The experimental data showed that the time-dependent release of the three fractions from soy protein gels fit Fick's second law. An increasing protein concentration or heating time resulted in a decrease in diffusion coefficients of non-network proteins. A power law expression was used to describe the relationship between non-network protein diffusion coefficient and molecular weight, for which the exponent (α) shifted to higher value with an increase in protein concentration or heating time, indicating that a more compact gel structure was formed.