EMAIL THIS PAGE TO A FRIEND
Behavioural Brain Research
Distribution of serotonin 5-HT1A-binding sites in the brainstem and the hypothalamus, and their roles in 5-HT-induced sleep and ingestive behaviors in rock pigeons (Columba livia).
PMID 25843559