EMAIL THIS PAGE TO A FRIEND
Cardiovascular Diabetology
Poorly controlled type 2 diabetes is accompanied by significant morphological and ultrastructural changes in both erythrocytes and in thrombin-generated fibrin: implications for diagnostics.
PMID 25848817