PloS one
Sex differences in liver toxicity-do female and male human primary hepatocytes react differently to toxicants in vitro?
PMID 25849576
Abstract
There is increasing amount of evidence for sex variation in drug efficiency and toxicity profiles. Women are more susceptible than men to acute liver injury from xenobiotics. In general, this is attributed to sex differences at a physiological level as well as differences in pharmacokinetics and pharmacodynamics, but neither of these can give a sufficient explanation for the diverse responses to xenobiotics. Existing data are mainly based on animal models and limited data exist on in vitro sex differences relevant to humans. To date, male and female human hepatocytes have not yet been compared in terms of their responses to hepatotoxic drugs. We investigated whether sex-specific differences in acute hepatotoxicity can be observed in vitro by comparing hepatotoxic drug effects in male and female primary human hepatocytes. Significant sex-related differences were found for certain parameters and individual drugs, showing an overall higher sensitivity of female primary hepatocytes to hepatotoxicants. Moreover, our work demonstrated that high content screening is feasible with pooled primary human hepatocytes in suspension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V4629
(±)-Verapamil hydrochloride, ≥99% (titration), powder
C27H38N2O4 · HCl
PHR1131
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C27H38N2O4 · HCl
C8138
Chlorpromazine hydrochloride, ≥98% (TLC)
C17H19ClN2S · HCl
C0982
Chlorpromazine hydrochloride, meets USP testing specifications
C17H19ClN2S · HCl
C1900000
Chlorpromazine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C17H19ClN2S · HCl
1125006
Chlorpromazine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C17H19ClN2S · HCl
31679
Chlorpromazine hydrochloride, VETRANAL, analytical standard
C17H19ClN2S · HCl
Y0001635
Diclofenac for system suitability, European Pharmacopoeia (EP) Reference Standard
C14H10Cl2NNaO2
S0765000
Diclofenac sodium, European Pharmacopoeia (EP) Reference Standard
C14H10Cl2NNaO2
1188800
Diclofenac sodium, United States Pharmacopeia (USP) Reference Standard
C14H10Cl2NNaO2
D6899
Diclofenac sodium salt
C14H10Cl2NNaO2
93484
Diclofenac sodium salt, analytical standard
C14H10Cl2NNaO2
PHR1144
Diclofenac sodium salt, Pharmaceutical Secondary Standard; Certified Reference Material
C14H10Cl2NNaO2
O104
Omeprazole, solid
C17H19N3O3S
19329
Omeprazole, analytical standard
C17H19N3O3S
PHR1059
Omeprazole, Pharmaceutical Secondary Standard; Certified Reference Material
C17H19N3O3S
1478505
Omeprazole, United States Pharmacopeia (USP) Reference Standard
C17H19N3O3S
Y0001042
Omeprazole for peak identification, European Pharmacopoeia (EP) Reference Standard
C17H19N3O3S
1711202
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard
C27H38N2O4 · HCl
V0100000
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
C27H38N2O4 · HCl