EMAIL THIS PAGE TO A FRIEND
Acta Poloniae Pharmaceutica
Synthesis and evaluation of anticonvulsant activity of N-(2,5-dimethylphenoxy)- and N-[(2,3,5-trimethylphenoxy)alkyl]aminoalkanols.
PMID 25850204