EMAIL THIS PAGE TO A FRIEND
AMB Express
Microbial production of Propionic and Succinic acid from Sorbitol using Propionibacterium acidipropionici.
PMID 25852990
Abstract
Three sequential fermentative batches were carried out with cell recycle in four simultaneously operating bioreactors maintained at pHxa06.5, 30°C, and 100xa0rpm. P. acidipropionici ATCC 4875 was able to produce propionic and succinic acid from sorbitol. The concentration of propionic acid decreased slightly from 39.5 ± 5.2xa0gxa0L(-1) to 34.4 ± 1.9xa0gxa0L(-1), and that of succinic acid increased significantly from 6.1 ± 2.1xa0gxa0L(-1) to 14.8 ± 0.9xa0gxa0L(-1) through the sequential batches. In addition, a small amount of acetic acid was produced that decreased from 3.3 ± 0.4xa0gxa0L(-1) to 2.0 ± 0.3xa0gxa0L(-1) through the batches. The major yield for propionic acid was 0.613xa0gxa0g(-1) in the first batch and succinic acid it was 0.212xa0gxa0g(-1) in the third batch. The minor yield of acetic acid was 0.029xa0gxa0g(-1), in the second and third batches.