EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Cloning, purification and evaluation of the enzymatic properties of a novel arylacetonitrilase from Luminiphilus syltensis NOR5-1B: a potential biocatalyst for the synthesis of mandelic acid and its derivatives.
PMID 25854992
Abstract
To examine nitrilase-mediated hydrolysis of nitriles to produce optically pure α-hydroxycarboxylic acids. A novel nitrilase, GPnor51, from Luminiphilus syltensis NOR5-1B was discovered by genomic data mining. It could hydrolyze racemic o-chloromandelonitrile to (R)-o-chloromandelic acid with high enantioselectivity (ee 98.2 %). GPnor51 was overexpressed in Escherichia coli BL21 (DE3), purified, and its catalytic properties studied. GPnor51 had a broad substrate acceptance toward various nitriles with structure diversity. It was an arylacetonitrilase that uses arylacetonitriles as optimal substrates. The V max and K m of GPnor51 towards o-chloromandelonitrile were 1.9 μmol min(-1) mg(-1) protein and 0.38 mM, respectively. GPnor51 also demonstrated high enantioselectivity toward mandelonitrile and other substituted mandelonitrile. This enzyme has a great potential for commercial production of optically pure (R)-mandelic acid and its derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2625
DL-Mandelic acid
C8H8O3
M2101
Mandelic acid, 99%
C8H8O3