EMAIL THIS PAGE TO A FRIEND
BMC medicine
Proxy markers of serum retinol concentration, used alone and in combination, to assess population vitamin A status in Kenyan children: a cross-sectional study.
PMID 25856672
Abstract
Serum retinol concentration determined by high-performance liquid chromatography (HPLC) is recommended by the World Health Organization to assess population vitamin A status. This assay is expensive, technically demanding and rarely available in developing countries. Our objective was a) to assess the diagnostic performance of proxy markers in detecting vitamin A deficiency and b) to derive decision rules based on these markers to estimate vitamin A deficiency prevalence. A survey was conducted in 15 rural primary schools in Eastern Province, Kenya, with 375 children aged 6 to 12xa0years (25 randomly selected per school). Serum retinol concentration <0.70xa0μmol/L by HPLC was used to define vitamin A deficiency. Proxy markers for vitamin A deficiency were serum concentrations of retinol binding protein (RBP), transthyretin, retinol measured by fluorometry and RBP:transthyretin molar ratio. The prevalence of vitamin A deficiency (HPLC) was 18%. Transthyretin and RBP showed the best diagnostic performance individually, with area-under-the-curve (AUC) values of 0.96 and 0.93. When combined, and with C-reactive protein added, the AUC increased to 0.98. A simple decision rule {(-15.277 × [RBP, μmol/L] - 7.013 × [Transthyretin, μmol/L] + 0.367 × [C-reactive protein, mg/L] + 24.714) > 0.496} yielded prevalence estimates of vitamin A deficiency that is unbiased by diagnostic error. The combination of transthyretin, RBP and C-reactive protein concentrations could eventually replace retinol concentration by HPLC in resource-poor settings as the preferred method to assess the population burden of vitamin A deficiency.