EMAIL THIS PAGE TO A FRIEND
Journal of perinatology : official journal of the California Perinatal Association
Predictors of successful closure of patent ductus arteriosus with indomethacin.
PMID 25856764
Abstract
To determine whether platelet counts can predict the likelihood of successful closure of patent ductus arteriosus (PDA) with indomethacin. This was a retrospective cohort study of infants <32 weeks' gestational age (GA) and birth weight <1500 g with PDA. Clinical characteristics between infants who achieved ductal closure with indomethacin and those who failed were compared. Multivariable logistic regression was used to identify predictors of successful ductal closure. In infants with hemodynamically significant PDA, older GA (odds ratio=1.54; 95% confidence interval: 1.12 to 2.13), male gender (odds ratio=3.02; 95% confidence interval: 1.08 to 8.49) and higher platelet count (odds ratio=1.5; 95% confidence interval: 1.04 to 2.17) prior to indomethacin treatment were associated with successful ductal closure with indomethacin. Older GA, male gender and higher platelet count at time of treatment of hemodynamically significant PDA are predictors of successful ductal closure with indomethacin.