EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Chloride channels mediate sodium sulphide-induced relaxation in rat uteri.
PMID 25857480
Abstract
Hydrogen sulphide reduces uterine contractility and is of potential interest as a treatment for uterine disorders. The aim of this study was to explore the mechanism of sodium sulphide (Na2 S)-induced relaxation of rat uterus, investigate the importance of redox effects and ion channel-mediated mechanisms, and any interactions between these two mechanisms. Organ bath studies were employed to assess the pharmacological effects of Na2 S in uterine strips by exposing them to Na2 S with or without Cl(-) channel blockers (DIDS, NFA, IAA-94, T16Ainh-A01, TA), raised KCl (15 and 75 mM), K(+) channel inhibitors (glibenclamide, TEA, 4-AP), L-type Ca(2+) channel activator (S-Bay K 8644), propranolol and methylene blue. The activities of antioxidant enzymes were measured in homogenates of treated uteri. The expression of bestrophin channel 1 (BEST-1) was determined by Western blotting and RT-PCR. Na2 S caused concentration-dependent reversible relaxation of spontaneously active and calcium-treated uteri, affecting both amplitude and frequency of contractions. Uteri exposed to 75 mM KCl were less sensitive to Na2 S compared with uteri in 15 mM KCl. Na2 S-induced relaxations were abolished by DIDS, but unaffected by other modulators or by the absence of extracellular HCO3 (-) , suggesting the involvement of chloride ion channels. Na2 S in combination with different modulators provoked specific changes in the anti-oxidant profiles of uteri. The expression of BEST-1, both mRNA and protein, was demonstrated in rat uteri. The relaxant effects of Na2 S in rat uteri are mediated mainly via a DIDS-sensitive Cl(-) -pathway. Components of the relaxation are redox- and Ca(2+) -dependent.