EMAIL THIS PAGE TO A FRIEND
The International Journal of Tuberculosis and Lung Disease : the Official Journal of the International Union Against Tuberculosis and Lung Disease
Genetic mutations associated with rifampicin and isoniazid resistance in MDR-TB patients in North-West India.
PMID 25859999