EMAIL THIS PAGE TO A FRIEND
Leukemia & Lymphoma
The novel kinesin spindle protein (KSP) inhibitor SB-743921 exhibits marked activity in in vivo and in vitro models of aggressive large B-cell lymphoma.
PMID 25860245