EMAIL THIS PAGE TO A FRIEND
Anticancer research
Dual expression of shAkt1 and Pdcd4 suppresses lung tumorigenesis in K-rasLA1 mice.
PMID 25862855
Abstract
Lung cancer has the highest mortality rate among cancers and current therapies are not efficient. Therefore, novel therapeutic methods are urgently needed. Here, we examined the effectiveness of simultaneous Akt1 inhibition and Pdcd4 over-expression using a dual expression system in suppressing tumorigenesis in K-ras(LA1) mice (a lung cancer model). An shRNA targeting Akt1 (shAkt1) and cDNA of programmed cell death protein 4 (Pdcd4) were inserted into a dual expression vector (shAkt1+Pdcd4). A sorbitol diacrylate-polyethylenimine (SDA-PEI) carrier was used because of low toxicity and high transfection efficiency. Aerosolized SDA-PEI/shAkt1+Pdcd4 complex was delivered to the mice twice a week for 4 weeks using a nose-only exposure inhalation chamber. Simultaneous Akt1 inhibition and Pdcd4 over-expression synergistically induced potent antitumor effect. Analysis revealed significant reduction in lung tumor number. Dual expression of shAkt1 and Pdcd4 effectively suppresses lung tumorigenesis.