EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Characterization of the impurities and isomers in cefetamet pivoxil hydrochloride by liquid chromatography/time-of-flight mass spectrometry and ion trap mass spectrometry.
PMID 25863019
Abstract
Ten impurities and isomers in cefetamet pivoxil hydrochloride drug substance made in China were separated and identified by HPLC-MS(n) (TOF and TRAP). Their fragmentation patterns and structural assignment were studied based on the HPLC-MS(n) data. Among the ten impurities and isomers, impurity VII was isolated by preparative HPLC, and its structure was confirmed by (1)H NMR data.