EMAIL THIS PAGE TO A FRIEND
Journal of cardiothoracic and vascular anesthesia
Viscoelastic blood coagulation measurement with Sonoclot predicts postoperative bleeding in cardiac surgery after heparin reversal.
PMID 25864405
Abstract
The aim of the study was to determine if Sonoclot with its sensitive glass bead-activated, viscoelastic test can predict postoperative bleeding in patients undergoing cardiac surgery at predefined time points. A prospective, observational clinical study. A teaching hospital, single center. Consecutive patients undergoing cardiac surgery (N = 300). Besides routine laboratory coagulation studies and heparin management with standard (kaolin) activated clotting time, additional native blood samples were analyzed on a Sonoclot using glass bead-activated tests. Glass bead-activated clotting time, clot rate, and platelet function were recorded immediately before anesthesia induction and at the end of surgery after heparin reversal but before chest closure. Primary outcome was postoperative blood loss (chest tube drainage at 4, 8, and 12 hours postoperatively). Secondary outcome parameters were transfusion requirements, need for surgical re-exploration, time of mechanical ventilation, length of intensive care unit and hospital stay, and hospital morbidity and mortality. Patients were categorized into "bleeders" and "nonbleeders." Patient characteristics, operations, preoperative standard laboratory parameters, and procedural times were comparable between bleeders and nonbleeders except for sex and age. Bleeders had higher rates of transfusions, surgical re-explorations, and complications. Only glass bead measurements by Sonoclot after heparin reversal before chest closure but not preoperatively were predictive for increased postoperative bleeding. Sonoclot with its glass bead-activated tests may predict the risk for postoperative bleeding in patients undergoing cardiac surgery at the end of surgery after heparin reversal but before chest closure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
857653
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
C8H15NO2